Learn More
Desmethyl Sibutramine, Hydrochloride Salt, TRC
Product Code. 30137867
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137867 5 mg 5mg
30135479 10 mg 10mg
30012222 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30137867 Supplier Toronto Research Chemicals Supplier No. TRCD2921705MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 84467-94-7
Molecular Formula C16 H24 Cl N . Cl H
Molecular Weight (g/mol) 302.28
Synonym N-Monodesmethylsibutramine hydrochloride, Cyclobutanemethanamine, 1-(4-chlorophenyl)-N-methyl-α-(2-methylpropyl)-, hydrochloride (1:1), Cyclobutanemethanamine, 1-(4-chlorophenyl)-N-methyl-α-(2-methylpropyl)-, hydrochloride (9CI), BTS 54-354, Desmethylsibutramine hydrochloride, (1RS)-1-[1-(4-Chlorophenyl)cyclobutyl]-N,3-dimethylbutan-1-amine hydrochloride
IUPAC Name 1-[1-(4-chlorophenyl)cyclobutyl]-N,3-dimethylbutan-1-amine;hydrochloride
SMILES Cl.CNC(CC(C)C)C1(CCC1)c2ccc(Cl)cc2
Specifications
Quantity 5 mg
InChI Formula InChI=1S/C16H24ClN.ClH/c1-12(2)11-15(18-3)16(9-4-10-16)13-5-7-14(17)8-6-13;/h5-8,12,15,18H,4,9-11H2,1-3H3;1H
Formula Weight 301.1364
Percent Purity >95%
Chemical Name or Material Desmethylsibutramine Hydrochloride
Grade HPLC