missing translation for 'onlineSavingsMsg'
Learn More

Desmethyl Doxylamine, TRC

Product Code. 30135621
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135621 1 g 1g
30007562 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135621 Supplier Toronto Research Chemicals Supplier No. TRCD2915251G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1221-70-1
Molecular Formula C16H20N2O
Molecular Weight (g/mol) 256.34
Synonym Ethanamine, N,N-dimethyl-2-(phenyl-2-pyridinylmethoxy)-, Pyridine, 2-[α-[2-(dimethylamino)ethoxy]benzyl]- (6CI,7CI), Pyridine, 2-[α-[2-(trimethylamino)ethoxy]benzyl]- (8CI), N,N-Dimethyl-2-(phenyl-2-pyridinylmethoxy)ethanamine, N,N-Dimethyl-2-[(RS)-1-phenyl(pyridin-2-yl)methoxy]ethanamine
SMILES CN(C)CCOC(c1ccccc1)c2ccccn2

Specifications

Quantity 1 g
InChI Formula InChI=1S/C16H20N2O/c1-18(2)12-13-19-16(14-8-4-3-5-9-14)15-10-6-7-11-17-15/h3-11,16H,12-13H2,1-2H3
Formula Weight 256.1576
Percent Purity >95%
Chemical Name or Material Desmethyl Doxylamine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.