missing translation for 'onlineSavingsMsg'
Learn More

Desethyldesisopropyl Atrazine, TRC

Product Code. 30008253
Change view
Click to view available options
Quantity:
500 mg
5 g
Unit Size:
500mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008253 5 g 5g
17933525 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30008253 Supplier Toronto Research Chemicals Supplier No. TRCD2891005G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3397-62-4
Molecular Formula C3 H4 Cl N5
Molecular Weight (g/mol) 145.55
Synonym s-Triazine, 2,4-diamino-6-chloro- (6CI,7CI,8CI), 6-Chloro-1,3,5-triazine-2,4-diamine, 2,4-Diamino-6-chloro-1,3,5-triazine, 2,4-Diamino-6-chloro-s-triazine, 2,4-Diamino-6-chlorotriazine, 2-Chloro-1,3,5-triazine-4,6-diamine, 2-Chloro-4,6-bis(amino)-1,3,5-triazine, 2-Chloro-4,6-bisamino-s-triazine, 2-Chloro-4,6-diamino-1,3,5-triazine, 2-Chloro-4,6-diamino-s-triazine, 4,6-Diamino-2-chloro-s-triazine, 6-Chloro-2,4-diamino-1,3,5-triazine, AtraMod, Chloroammeline, DACT, Deethyldeisopropylatrazine, Deisopropyldeethyl atrazine, Desethyldesisopropylatrazine, Didealkylatrazine, Dideethylsimazine, ENT 50982, G 28273, NSC 680830, NSC 7965
IUPAC Name 6-chloro-1,3,5-triazine-2,4-diamine
SMILES Nc1nc(N)nc(Cl)n1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C3H4ClN5/c4-1-7-2(5)9-3(6)8-1/h(H4,5,6,7,8,9)
Formula Weight 145.0155
Chemical Name or Material Atrazine-desethyl-desisopropyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.