missing translation for 'onlineSavingsMsg'
Learn More

Desethyl Dabigatran Etexilate, TRC

Product Code. 30352702
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30352702 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30352702 Supplier Toronto Research Chemicals Supplier No. TRCD28907050MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Desethyldabigatran Etexilate
CAS 212321-78-3
Purity Grade Notes HPLC
Molecular Formula C32 H37 N7 O5
Quantity 50 mg
Synonym Desethyl Dabigatran Etexilate, BIBR 1087 SE, N-[[2-[[[4-[[[(Hexyloxy)carbonyl]amino]iminomethyl]phenyl]amino]methyl]-1-methyl-1H-benzimidazol-5-yl]carbonyl]-N-2-pyridinyl-β-alanine
InChI Formula InChI=1S/C32H37N7O5/c1-3-4-5-8-19-44-32(43)37-30(33)22-10-13-24(14-11-22)35-21-28-36-25-20-23(12-15-26(25)38(28)2)31(42)39(18-16-29(40)41)27-9-6-7-17-34-27/h6-7,9-15,17,20,35H,3-5,8,16,18-19,21H2,1-2H3,(H,40,41)(H2,33,37,43)
SMILES CCCCCCOC(=O)NC(=N)c1ccc(NCc2nc3cc(ccc3n2C)C(=O)N(CCC(=O)O)c4ccccn4)cc1
IUPAC Name 3-[[2-[[4-(N-hexoxycarbonylcarbamimidoyl)anilino]methyl]-1-methylbenzimidazole-5-carbonyl]-pyridin-2-ylamino]propanoic acid
Molecular Weight (g/mol) 599.68
Formula Weight 599.2856
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.