Learn More
Deoxyarbutin, TRC
Product Code. 30354436
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30354436 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30354436 Supplier Toronto Research Chemicals Supplier No. TRCD23210550MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
CAS 53936-56-4
Molecular Formula C11H14O3
Quantity 50 mg
Synonym Deoxyarbutin, Phenol, 4-[(tetrahydro-2H-pyran-2-yl)oxy]-, 4-[(Tetrahydro-2H-pyran-2-yl)oxy]phenol, 4-(Tetrahydro-2H-pyran-2-yloxy)phenol, 4-Hydroxyphenyl tetrahydropyranyl ether, 4-[(2-Tetrahydropyranyl)oxy]phenol, Deoxyarbutin, 4-(Tetrahydro-2H-pyran-2-yloxy)phenol, 4-Hydroxyphenyl tetrahydropyranyl Ether, 4-[(2-Tetrahydropyranyl)oxy]phenol
InChI Formula InChI=1S/C11H14O3/c12-9-4-6-10(7-5-9)14-11-3-1-2-8-13-11/h4-7,11-12H,1-3,8H2
SMILES Oc1ccc(OC2CCCCO2)cc1
IUPAC Name 4-(oxan-2-yloxy)phenol
Molecular Weight (g/mol) 194.23
Formula Weight 194.0943
Chemical Name or Material Deoxyarbutin