Learn More
Emma Safety Footwear Dempo Safety Shoes
Product Code. 15968791
Change view
Click to view available options
Size:
37
38
39
40
41
42
43
44
45
46
47
48
49
Unit Size:
1 pair
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15968791 41 1 pair
15928791 37 1 pair
15938791 38 1 pair
15948791 39 1 pair
15958791 40 1 pair
15978791 42 1 pair
15988791 43 1 pair
15998791 44 1 pair
15908801 45 1 pair
15918801 46 1 pair
15928801 47 1 pair
15938801 48 1 pair
15948801 49 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Product Code. 15968791 Supplier Emma Safety Footwear Supplier No. MM58384841
  • ONLINE EXCLUSIVE PRICE
    203.52 EUR
    119.70 EUR / 1 pairs
    Save 83.82 EUR
    41% Off
Estimated Shipment: 12-04-2027
Extreme safety in extreme temperatures. Easy on/off slip-on lightweight leather boot with a double density rubber-PU sole. The Nitrile rubber outsole repels corrosive chemicals and substances, even liquid, molten metals. Heat resistant up to 300°C.
  • S3-Safety shoe with sole plate for wet work
  • Full grain leather construction
  • GripForce™ Duo Heat Resistant Outsole
  • Sanitized
Specifications
Certifications/Compliance EN ISO 20345:2011
Color Black
Design Dempo
Liner Material Hydro-Tec™ Sanitized Silver lined tongue
Material Leather
Size 41
Toe Material Steel