missing translation for 'onlineSavingsMsg'
Learn More

Demeton-S, TRC

Product Code. 18062040
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18062040 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18062040 Supplier Toronto Research Chemicals Supplier No. TRCD231360100MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 126-75-0
Molecular Formula C8 H19 O3 P S2
Quantity 100 mg
Synonym Demeton thiolo, Demeton-S, Demetonthiol, Ethylthionodemeton, Isosystox, O,O-Diethyl S-[2-(ethylthio)ethyl] phosphorothioate, O,O-Diethyl S-[2-(ethylthio)ethyl] thiophosphate, Systox-thiol, Thioldemeton
InChI Formula InChI=1S/C8H19O3PS2/c1-4-10-12(9,11-5-2)14-8-7-13-6-3/h4-8H2,1-3H3
SMILES CCOP(=O)(OCC)SCCSCC
IUPAC Name 1-diethoxyphosphorylsulfanyl-2-ethylsulfanylethane
Molecular Weight (g/mol) 258.34
Formula Weight 258.0513
Chemical Name or Material Demeton-S
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.