Learn More
Demecarium Bromide, TRC
Product Code. 30363890
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30363890 10 mg 10mg
30360699 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30363890 Supplier Toronto Research Chemicals Supplier No. TRCD23072010MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 56-94-0
Molecular Formula C32 H52 N4 O4 . 2 Br
Molecular Weight (g/mol) 716.59
Synonym Benzenaminium, 3,3'-[1,10-decanediylbis[(methylimino)carbonyloxy]]bis[N,N,N-trimethyl-, bromide (1:2), (m-Hydroxyphenyl)trimethylammonium bromide decamethylenebis[methylcarbamate] (6CI,7CI), Ammonium, (m-hydroxyphenyl)trimethyl-, bromide, decamethylenebis[methylcarbamate] (2:1) (8CI), Benzenaminium, 3,3'-[1,10-decanediylbis[(methylimino)carbonyloxy]]bis[N,N,N-trimethyl-, dibromide (9CI), BC 48, Decamethylenebis(m-dimethylaminophenyl-N-methylcarbamate) dimethobromide, Decamethylenebis[N-methylcarbamic acid m-dimethylaminophenyl ester] bromomethylate, Demecarium bromide, Demekarium bromide, Demekastigmine bromide, Frumtosnil, Humorsol, N,N'-Bis[3-trimethylammoniumphenoxycarbonyl]-N,N'-dimethyldecamethylenediamine dibromide, Tonilen, Tosmicil, Tosmilen, Tosmilene, Visumatic, Visumiotic
IUPAC Name trimethyl-[3-[methyl-[10-[methyl-[3-(trimethylazaniumyl)phenoxy]carbonylamino]decyl]carbamoyl]oxyphenyl]azanium;dibromide
SMILES [Br-].[Br-].CN(CCCCCCCCCCN(C)C(=O)Oc1cccc(c1)[N+](C)(C)C)C(=O)Oc2cccc(c2)[N+](C)(C)C
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C32H52N4O4.2BrH/c1-33(31(37)39-29-21-17-19-27(25-29)35(3,4)5)23-15-13-11-9-10-12-14-16-24-34(2)32(38)40-30-22-18-20-28(26-30)36(6,7)8;;/h17-22,25-26H,9-16,23-24H2,1-8H3;2*1H/q+2;;/p-2
Formula Weight 714.2355
Chemical Name or Material Demecarium Bromide