missing translation for 'onlineSavingsMsg'
Learn More

Demecarium Bromide, Mikromol™

Product Code. 30049098
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30049098 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30049098 Supplier Mikromol Supplier No. MM3042.00
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Demecarium Bromide
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Demecarium Bromide
CAS 56-94-0
Analyte or Component Names Demecarium Bromide
Shipping Condition Room Temperature
Molecular Formula C32 H52 N4 O4 . 2 Br
Certifications/Compliance ISO 17025
Synonym Benzenaminium, 3,3'-[1,10-decanediylbis[(methylimino)carbonyloxy]]bis[N,N,N-trimethyl-, bromide (1:2), (m-Hydroxyphenyl)trimethylammonium bromide decamethylenebis[methylcarbamate] (6CI,7CI), Ammonium, (m-hydroxyphenyl)trimethyl-, bromide, decamethylenebis[methylcarbamate] (2:1) (8CI), Benzenaminium, 3,3'-[1,10-decanediylbis[(methylimino)carbonyloxy]]bis[N,N,N-trimethyl-, dibromide (9CI), BC 48, Decamethylenebis(m-dimethylaminophenyl-N-methylcarbamate) dimethobromide, Decamethylenebis[N-methylcarbamic acid m-dimethylaminophenyl ester] bromomethylate, Demecarium bromide, Demekarium bromide, Demekastigmine bromide, Frumtosnil, Humorsol, N,N'-Bis[3-trimethylammoniumphenoxycarbonyl]-N,N'-dimethyldecamethylenediamine dibromide, Tonilen, Tosmicil, Tosmilen, Tosmilene, Visumatic, Visumiotic
InChI Formula InChI=1S/C32H52N4O4.2BrH/c1-33(31(37)39-29-21-17-19-27(25-29)35(3,4)5)23-15-13-11-9-10-12-14-16-24-34(2)32(38)40-30-22-18-20-28(26-30)36(6,7)8;;/h17-22,25-26H,9-16,23-24H2,1-8H3;2*1H/q+2;;/p-2
SMILES [Br-].[Br-].CN(CCCCCCCCCCN(C)C(=O)Oc1cccc(c1)[N+](C)(C)C)C(=O)Oc2cccc(c2)[N+](C)(C)C
IUPAC Name trimethyl-[3-[methyl-[10-[methyl-[3-(trimethylazaniumyl)phenoxy]carbonylamino]decyl]carbamoyl]oxyphenyl]azanium;dibromide
Molecular Weight (g/mol) 716.59
Quantity 100 mg
Formula Weight 714.2355 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.