missing translation for 'onlineSavingsMsg'
Learn More

Demecarium Bromide, MedChemExpress

Product Code. 16463806
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16463806 25 mg 25mg
16421128 5 mg 5mg
16418718 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16463806 Supplier Medchem Express Supplier No. HYB1626A
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Demecarium Bromide (BC-48) is a potent cholinesterase inhibitor, with an apparent affinity (Kiapp) of 0.15 μM. Demecarium Bromide (BC-48) is used as a glaucoma agent.

Demecarium Bromide (BC-48) is a potent cholinesterase inhibitor, with an apparent affinity (Kiapp) of 0.15 μM. Demecarium Bromide (BC-48) is used as a glaucoma agent.

Specifications

Chemical Name or Material Demecarium Bromide
CAS 56-94-0
Color White
Purity Grade Notes Research
Molecular Formula C32H52Br2N4O4
Quantity 25 mg
Synonym BC-48
Solubility Information DMSO : 160 mg/mL (223.28 mM; Need ultrasonic)
SMILES O=C(N(CCCCCCCCCCN(C(OC1=CC([N+](C)(C)C)=CC=C1)=O)C)C)OC2=CC([N+](C)(C)C)=CC=C2.[Br-].[Br-]
Molecular Weight (g/mol) 716.59
Formula Weight 716.59
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.