missing translation for 'onlineSavingsMsg'
Learn More

Deltasonamide 2 (TFA), MedChemExpress

Product Code. p-200102139
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16475786 100 mg 100mg
16455048 10 mg 10mg
16421287 50 mg 50mg
16407777 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16475786 Supplier Medchem Express Supplier No. HY122641B
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Deltasonamide 2 TFA is competitive, high affinity PDEδ inhibitor with a Kd of ∼385 pM.

Deltasonamide 2 TFA is competitive, high affinity PDEδ inhibitor with a Kd of ∼385 pM.

Specifications

Chemical Name or Material Deltasonamide 2 (TFA)
CAS 2235358-74-2
Color White
Purity Grade Notes Research
Molecular Formula C32H40ClF3N6O6S2
Quantity 100 mg
Solubility Information DMSO : 125 mg/mL (164.20 mM; Need ultrasonic)
SMILES O=S(C1=CC=C(S(=O)(N(CC2=CC=C(Cl)C=C2)C3CCCC3)=O)C=C1)(N([C@H]4CC[C@H](N)CC4)CC5=NC(NC)=NC=C5)=O.O=C(O)C(F)(F)F
Molecular Weight (g/mol) 761.27
Formula Weight 761.27
Percent Purity 99.65%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.