missing translation for 'onlineSavingsMsg'
Learn More

Delamanid, MedChemExpress

Product Code. 16464198
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16464198 10 mg 10mg
16490126 2 mg 2mg
16472986 25 mg 25mg
16465156 100 mg 100mg
16436917 5 mg 5mg
16430547 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16464198 Supplier Medchem Express Supplier No. HY10846
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Delamanid, a newer mycobacterial cell wall synthesis inhibitor, inhibits the synthesisi of mucolic acids.

Delamanid, a newer mycobacterial cell wall synthesis inhibitor, inhibits the synthesisi of mucolic acids.

Specifications

Chemical Name or Material Delamanid
CAS 681492-22-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C25H25F3N4O6
Quantity 10 mg
Synonym OPC-67683
Solubility Information DMSO : ≥ 50 mg/mL (93.55 mM) ∣Ethanol : 2 mg/mL (3.74 mM; Need ultrasonic)
SMILES FC(F)(F)OC1=CC=C(OC2CCN(C3=CC=C(OC[C@@]4(C)CN5C(O4)=NC([N+]([O-])=O)=C5)C=C3)CC2)C=C1
Molecular Weight (g/mol) 534.48
Formula Weight 534.48
Percent Purity 98.72%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.