Learn More
Elgas™ Deioniser Vision 125
Product Code. 13145940
Change view
Click to view available options
Height (Metric):
385 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Unit Size
13145940 385 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13145940 Supplier Elgas™ Supplier No. PRDIDC0800
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
The Vision range of wall mounted, disposable deionisers produce deionised water at the turn of a tap
The Vision range of wall mounted, disposable deionisers produce deionised water at the turn of a tap. The use of colour change resin, which indicates when it is exhausted, means that no electrical supply is needed to operate these deionisers. Deionised water is produced at up to 60L/hr. To help with dispensing the purified water, these disposable deionisers are also available complete with a flexible dispensing gun.
Specifications
Frequency 50/60 Hz
Height (Metric) 385 mm
Type Deioniser Vision 125
Voltage 230 V