Learn More
Dehydronitrosonisoldipine, TRC
Product Code. 30365851
Change view
Click to view available options
Quantity:
5 mg
50 mg
100 mg
Unit Size:
100mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30365851 100 mg 100mg
30362856 50 mg 50mg
30361703 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30365851 Supplier Toronto Research Chemicals Supplier No. TRCD230310100MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 87375-91-5
Molecular Formula C20H22N2O5
Molecular Weight (g/mol) 370.4
Synonym 2,6-Dimethyl-4-(2-nitrosophenyl)-3,5-pyridinedicarboxylic Acid 3-Methyl 5-(2-Methylpropyl) Ester, 2,6-Dimethyl-4-(2-nitrosophenyl)-3,5-pyridinedicarboxylic Acid Methyl 2-Methylpropyl Ester, 5-Methyl 3-(2-Methylprop-3-yl) 2,6-Dimethyl-4-(2-nitrosophenyl)pyridine-3,5-dicarboxylate, Nisoldipine Impurity D
SMILES COC(=O)c1c(C)nc(C)c(C(=O)OCC(C)C)c1c2ccccc2N=O
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C20H22N2O5/c1-10(2)13(5)27-20(25)17-12(4)21-11(3)16(19(23)24)18(17)14-8-6-7-9-15(14)22-26/h6-10,13H,1-5H3,(H,23,24)/p-1
Formula Weight 370.1529
Chemical Name or Material Dehydronitrosonisoldipine
Product Title
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.