missing translation for 'onlineSavingsMsg'
Learn More

Dehydronitrosonisoldipine, MedChemExpress

Product Code. 16470489
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16470489 10 mg 10mg
16452728 5 mg 5mg
16434136 25 mg 25mg
16413479 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16470489 Supplier Medchem Express Supplier No. HYZ0816
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Dehydronitrosonisoldipine, a derivative of Nisoldipine (HY-17402), is an irreversible and cell-permeant sterile alpha and TIR motif-containing 1 (SARM1) inhibitor.

Dehydronitrosonisoldipine acts mainly by blocking SARM1 activation but not its enzymatic activities. Dehydronitrosonisoldipine inhibits SARM1 and axon degenration (AxD) by covalently modifying cysteines, also inhibits the Vincristine-activated cADPR production in neurons. Dehydronitrosonisoldipine can be used for researching neurodegenerative disorders.

Specifications

Chemical Name or Material Dehydronitrosonisoldipine
CAS 87375-91-5
Color Green
Purity Grade Notes Research
Molecular Formula C20H22N2O5
Quantity 10 mg
Solubility Information DMSO : 100 mg/mL (269.98 mM; Need ultrasonic)
SMILES O=C(C1=C(C2=CC=CC=C2N=O)C(C(OCC(C)C)=O)=C(C)N=C1C)OC
Molecular Weight (g/mol) 370.4
Formula Weight 370.4
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.