missing translation for 'onlineSavingsMsg'
Learn More

Deferoxamine Mesylate, TRC

Product Code. 30134359
Change view
Click to view available options
Quantity:
25 mg
250 mg
1 g
Unit Size:
1g
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134359 25 mg 25mg
30133850 250 mg 250mg
17947814 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134359 Supplier Toronto Research Chemicals Supplier No. TRCD22898025MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138-14-7
Molecular Formula C25 H48 N6 O8 . C H4 O3 S
Molecular Weight (g/mol) 656.79
Synonym Butanediamide, N4-[5-[[4-[[5-(acetylhydroxyamino)pentyl]amino]-1,4-dioxobutyl]hydroxyamino]pentyl]-N1-(5-aminopentyl)-N1-hydroxy-, methanesulfonate (1:1), Deferoxamine mesilate, N'-[5-[[4-[[5-(Acetylhydroxyamino)pentyl]amino]-4-oxobutanoyl]hydroxyamino]pentyl]-N-(5-aminopentyl)-N-hydroxybutanediamide methanesulfonate, Butanediamide, N'-[5-[[4-[[5-(acetylhydroxyamino)pentyl]amino]-1,4-dioxobutyl]hydroxyamino]pentyl]-N-(5-aminopentyl)-N-hydroxy-, monomethanesulfonate (salt), Propionohydroxamic acid, N-[5-[3-[(5-aminopentyl)hydroxycarbamoyl]propionamido]pentyl]-3-[[5-(N-hydroxyacetamido)pentyl]carbamoyl]-, methanesulfonate, Propionohydroxamic acid, N-[5-[3-[(5-aminopentyl)hydroxycarbamoyl]propionamido]pentyl]-3-[[5-(N-hydroxyacetamido)pentyl]carbamoyl]-, monomethanesulfonate (salt), Ba 33112, DFOM, DFX mesylate, Deferoxamine B mesylate, Deferoxamine mesylate, Deferoxamine methanesulfonate, Deferrioxamine B methanesulfonate, Deferrioxamine methanesulfonate, Desferal, Desferal mesylate, Desferrioxamine B mesylate, Desferrioxamine B methanesulfonate, Desferrioxamine mesylate, Desferrioxamine methanesulfonate, NSC 644468
IUPAC Name N-[5-[[4-[5-[acetyl(hydroxy)amino]pentylamino]-4-oxobutanoyl]-hydroxyamino]pentyl]-N'-(5-aminopentyl)-N'-hydroxybutanediamide;methanesulfonic acid
SMILES CC(=O)N(O)CCCCCNC(=O)CCC(=O)N(O)CCCCCNC(=O)CCC(=O)N(O)CCCCCN.CS(=O)(=O)O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C25H48N6O8.CH4O3S/c1-21(32)29(37)18-9-3-6-16-27-22(33)12-14-25(36)31(39)20-10-4-7-17-28-23(34)11-13-24(35)30(38)19-8-2-5-15-26;1-5(2,3)4/h37-39H,2-20,26H2,1H3,(H,27,33)(H,28,34);1H3,(H,2,3,4)
Formula Weight 656.3415
Chemical Name or Material Deferoxamine mesilate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.