Learn More
Tocris Bioscience™ Deferoxamine mesylate
GREENER_CHOICE
Iron chelator; also hypoxia mimetic
Brand:  Tocris Bioscience™ 5764/50
Encompass
Product Code. 15767096
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Deferoxamine mesylate is an iron chelator. Also hypoxia mimetic and inhibitor of ferroptosis. Has neuroprotective effects in animal models of stroke, traumatic brain injury and neurodegenerative diseases.
Specifications
Specifications
N4-[5-[[4-[[5-(Acetylhydroxyamino)pentyl]amino-1,4-dioxobutyl]hydroxyamino]pentyl]-N1-(5-aminopentyl)-N1-hydroxybutanediamide methanesulfonate
138-14-7