Learn More
Decyltrimethylammonium Chloride, TRC
Product Code. 30138112
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.5g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138112 500 mg 500mg
30132465 2.5 g 2.5g
30010272 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30138112 Supplier Toronto Research Chemicals Supplier No. TRCD525798500MG
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 10108-87-9
Molecular Formula C13H30N . Cl
Molecular Weight (g/mol) 235.84
Synonym 1-Decanaminium, N,N,N-trimethyl-, chloride (1:1) (ACI), 1-Decanaminium, N,N,N-trimethyl-, chloride (9CI), Ammonium, decyltrimethyl-, chloride (8CI), Decyltrimethylammonium chloride (7CI), Capryltrimethylammonium chloride, n-Decyltrimethylammonium chloride, Trimethylcaprylammonium chloride, Trimethyldecylammonium chloride, Decyl(trimethyl)ammonium chloride
SMILES [Cl-].CCCCCCCCCC[N+](C)(C)C
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C13H30N.ClH/c1-5-6-7-8-9-10-11-12-13-14(2,3)4;/h5-13H2,1-4H3;1H/q+1;/p-1
Formula Weight 235.2067
Chemical Name or Material Decyltrimethylammonium Chloride