Learn More
Borer Chemie™ deconex 25 ORGANACID
Product Code. 15410380
Change view
Click to view available options
Quantity:
1 x 5 kg
1 x 10 kg
1 x 25 kg
Unit Size:
10kg
25kg
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15410380 1 x 5 kg 5kg
17405112 1 x 10 kg 10kg
10014490 1 x 25 kg 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15410380 Supplier Borer Chemie™ Supplier No. 502100.00KK5R
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
Cleaner and neutralizer for laboratory washers. Shortens rinsing phases and reduces water consumption. deconex 25 ORGANACID is phosphate-free and suitable for environmental and water laboratories. Pre-rinsing agent against acid-soluble residues.
  • Neutralizer and cleaner for automated use
  • Ecological solution without phosphates and surfactants
  • Neutralizes carried-over alkaline residues, reduces rinsing phases
  • Reduces the consumption of rinsing water
  • Typical application concentration: 3 mL/L
  • Excellent material compatibility, also for aluminium
  • Can be rinsed off without leaving residues
  • Also as pre-rinsing agent against acid-soluble deposits
  • Technical support by the manufacturer
Specifications
Form Liquid
pH ∼2.6 (1% Solution)
Composition Organic acid
Concentration Concentrate
Container Type Canister
Content And Storage 5°C to 25°C
Quantity 1 x 5 kg
Volume (English) 1.1 gal.
Volume (Metric) 4.3 L
Weight (English) 11 lb.
Weight (Metric) 5 kg
Show More Show Less
compliance-icons
Product Identifier
  • deconex 25 ORGANACID
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: citric acid