Learn More
Borer Chemie™ deconex 11 UNIVERSAL
Product Code. 10370161
Change view
Click to view available options
Quantity:
1 x 5 kg
10 x 1 kg
1 x 10 kg
Unit Size:
10kg
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10370161 1 x 5 kg 5kg
17416492 10 x 1 kg 10kg
10422121 1 x 10 kg 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
You may only order up to the available stock. For more details call customer services on +358 9 8027 6280.
Product Code. 10370161 Supplier Borer Chemie™ Supplier No. 500100.00K05W
You may only order up to the available stock. For more details call customer services on +358 9 8027 6280.
Liquid cleaning concentrate for manual cleaning of laboratory glassware. Contains active chlorine. deconex 11 UNIVERSAL is suitable for use in immersion and ultrasonic baths. This cleaner is a full substitute for chromosulphuric acid.
  • Concentrate for manual cleaning
  • High yield, typical concentration: 20-50 mL/L
  • Rinses off without leaving residues
  • For consistent and flawless results
  • Contains active chlorine
  • Without EDTA and NTA
  • Technical support by the manufacturer
Specifications
Form Liquid
pH ∼11.6 (1% Solution)
Composition Alkalis, surfactants, inorganic chlorine carrier
Concentration Concentrate
Container Type Canister
Content And Storage 5°C to 25°C
Quantity 1 x 5 kg
Volume (English) 1.2 gal.
Volume (Metric) 4.5 L
Weight (English) 11 lb.
Weight (Metric) 5 kg
Show More Show Less
compliance-icons
Product Identifier
  • deconex 11 UNIVERSAL
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 1 B
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P234-Keep only in original container.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P406-Store in corrosive resistant/ container with a resistant inner liner.