Learn More
Borer Chemie™ deconex 11 UNIVERSAL eco
GREENER_CHOICE
Product Code. 31337540
Change view
Click to view available options
Volume (Metric):
4.5 L
8.9 L
10 x 0.9 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric)
31337540 4.5 L
31337539 10 x 0.9 L
31337541 8.9 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 31337540 Supplier Borer Chemie™ Supplier No. 514505.00K05W
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
Liquid concentrate for cleaning of laboratory utensils in immersion, ultrasonic baths and in pipette washers
  • Concentrate for manual cleaning
  • Ecological solution without phosphates
  • Chromosulfuric acid replacement
  • Without EDTA, NTA and chlorine
  • Highly productive, typical concentration: 10 to 20 mL/L
  • Pre-cleaned glassware can be further cleaned in a washer without intermediate rinsing
  • Rinses off without leaving residues
  • For consistent and error-free results
  • Extra wide cleaning spectrum
  • Hard water compatible
  • Technical support by the manufacturer
Specifications
Product Type Alkaline Cleaning Concentrate
Color-coded No
Form Concentrated Liquid
pH ∼12.0
Scent Unscented
Sterility Non-sterile
Composition Alkalis, Complexing Agents, Surfactants, Solubilizer
Concentration 1% Solution
Container Type Canister
Content And Storage 5°C to 25°C
Density 1.12 g/mL
Quantity 1 Ea.
Volume (Metric) 4.5 L
Weight (Metric) 5 kg
Show More Show Less
compliance-icons
Product Identifier
  • deconex 11 UNIVERSAL eco
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501-Dispose of contents/container to .