missing translation for 'onlineSavingsMsg'
Learn More

Dechlorane 602, TRC

Product Code. 30138273
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138273 50 mg 50mg
17924475 10 mg 10mg
17953395 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138273 Supplier Toronto Research Chemicals Supplier No. TRCD22620050MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31107-44-5
Molecular Formula C14 H4 Cl12 O
Molecular Weight (g/mol) 613.62
Synonym 1,4:6,9-Dimethanodibenzofuran, 1,2,3,4,6,7,8,9,10,10,11,11-dodecachloro-1,4,4a,5a,6,9,9a,9b-octahydro-, 1,2,3,4,6,7,8,9,10,10,11,11-Dodecachloro-1,4,4a,5a,6,9,9a,9b-octahydro-1,4:6,9-dimethanodibenzofuran, Dec 602, Dechlorane 602
IUPAC Name 1,4,5,6,7,11,12,13,14,14,15,15-dodecachloro-9-oxapentacyclo[9.2.1.14,7.02,10.03,8]pentadeca-5,12-diene
SMILES ClC1=C(Cl)[C@@]2(Cl)[C@@H]3[C@@H](O[C@H]4[C@@H]3[C@]5(Cl)C(=C(Cl)[C@@]4(Cl)C5(Cl)Cl)Cl)[C@]1(Cl)C2(Cl)Cl

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C14H4Cl12O/c15-3-5(17)11(21)7-1(9(3,19)13(11,23)24)2-8(27-7)12(22)6(18)4(16)10(2,20)14(12,25)26/h1-2,7-8H/t1-,2+,7+,8-,9-,10-,11-,12-/m0/s1
Formula Weight 607.6524
Percent Purity >95%
Chemical Name or Material Dechlorane 602
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.