Learn More
Decanoic Acid, Sigma-Aldrich™
Product Code. 15888700
Change view
Click to view available options
Quantity:
100 mL
1 L
Unit Size:
1 L
100mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15888700 1 L 1 L
15878700 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15888700 Supplier Sigma Aldrich Supplier No. 8021691000
 Restricted Item
Estimated Shipment: 20-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 334-48-5
Specifications
Melting Point 29°C to 32°C
Density 0.89 g/cm3 at 20°C
Quantity 1 L
Molecular Formula CH3(CH2)8COOH
MDL Number MFCD00004441
Synonym Decanoic acid, Capric acid, Decanoic acid
Molecular Weight (g/mol) 172.26
Grade Ungraded
Chemical Name or Material Decanoic Acid