Learn More
Decanal, Sigma-Aldrich™
Specifications
Specifications
| CAS | 112-31-2 |
| Melting Point | 7°C |
| Density | 0.83 g/cm3 at 20°C |
| Boiling Point | 207°C to 209°C/1013 hPa |
| Molecular Formula | CH3(CH2)8CHO |
| MDL Number | MFCD00007031 |
| Quantity | 100 mL |
| Synonym | Decanal, Capraldehyde, Decanal |
| Molecular Weight (g/mol) | 156.27 |
| Grade | Research |