missing translation for 'onlineSavingsMsg'
Learn More

Decamethonium Bromide, MedChemExpress

Product Code. 16458528
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16458528 10 g 10g
16421008 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16458528 Supplier Medchem Express Supplier No. HYB0570
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Decamethonium Bromide is a nicotinic AChR partial agonist and neuromuscular blocking agent.

Decamethonium Bromide is a nicotinic AChR partial agonist and neuromuscular blocking agent.

Specifications

Chemical Name or Material Decamethonium Bromide
Physical Form Solid
CAS 541-22-0
Color White
Quantity 10 g
Purity Grade Notes Research
Molecular Formula C16H38Br2N2
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : 83.33 mg/mL (199.22 mM; Need ultrasonic) ∣H2O : ≥ 50 mg/mL (119.53 mM)
SMILES C[N+](C)(C)CCCCCCCCCC[N+](C)(C)C.[Br-].[Br-]
Molecular Weight (g/mol) 418.29
Formula Weight 418.29
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.