missing translation for 'onlineSavingsMsg'
Learn More

Decaglycerin, TRC

Product Code. 30385243
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30385243 5 g 5g
30383205 1 g -
30378469 10 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30385243 Supplier Toronto Research Chemicals Supplier No. TRCD9041005G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9041-07-0
Molecular Formula C30H62O21
Molecular Weight (g/mol) 758.8
Synonym Decaglycerol-Desmodur N100-Sorbitan Tristearate Copolymer, Great Oil DE 1
SMILES C(C(COCC(COCC(COCC(COCC(COCC(COCC(COCC(COCC(COCC(CO)O)O)O)O)O)O)O)O)O)O)O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C30H62O21/c31-1-21(33)3-43-5-23(35)7-45-9-25(37)11-47-13-27(39)15-49-17-29(41)19-51-20-30(42)18-50-16-28(40)14-48-12-26(38)10-46-8-24(36)6-44-4-22(34)2-32/h21-42H,1-20H2
Chemical Name or Material Decaglycerin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.