missing translation for 'onlineSavingsMsg'
Learn More

Decaborane, TRC

Product Code. 30368638
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30368638 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30368638 Supplier Toronto Research Chemicals Supplier No. TRCD2005001G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Decaborane
CAS 17702-41-9
Quantity 1 g
Molecular Formula B10H14
Synonym Boron hydride (B10H14), Decaborane(14), Decaborane (B10H14), NSC 39828, Tetradecahydrodecaborane, nido-Decaborane(14)
InChI Formula InChI=1S/B10H14/c1-3-5-2-6-4(1)7(3)9(3,5)8(5,6)10(4,6)7/h3-10H,1-2H3
SMILES [BH]123([BH]456[BH]27([H][BH]7389)[H]6)[BH]%10%118[BH]%12%13%14[BH]%1014[BH]%12%155[BH]%13%16([H]%15)[BH]%11%149[H]%16
Molecular Weight (g/mol) 122.22
Formula Weight 124.2026
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.