missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ DDR1-IN-1 dihydrochloride
GREENER_CHOICE

Product Code. 18771779
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18771779

Brand: Tocris Bioscience™ 5077/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective DDR1 inhibitor

DDR1-IN-1 dihydrochloride is a selective discoidin domain receptor 1 (DDR1) tyrosine kinase inhibitor (IC50 values are 105 and 413 nM for DDR1 and DDR2 respectively). Inhibits integrin-induced DDR1 autophosphorylation in an osteosarcoma cell line.
TRUSTED_SUSTAINABILITY

Specifications

CAS 1780303-76-5
Quantity 50 mg
Product Type DDR1-IN-1 dihydrochloride
Purity >98%
Molecular Weight (g/mol) 625.51
Inhibitors DDR1-IN-1 dihydrochloride

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.