missing translation for 'onlineSavingsMsg'
Learn More

DDIT4 Antibody - BSA Free, Novus Biologicals™

Product Code. 18646580 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.02 mL
0.1 mL
Unit Size:
0.01mL
0.02mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18646580 0.02 mL 0.02mL
18683141 0.1 mL 0.01mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18646580 Supplier Novus Biologicals Supplier No. NBP2929420.02ml

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DDIT4 Polyclonal antibody specifically detects DDIT4 in Human samples. It is validated for Western Blot, Gene Knock-Out
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDIT4
Applications Western Blot, Gene Knock-Out
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:1000, Knockout Validated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias Dig2, DNA-damage-inducible transcript 4, FLJ20500, HIF-1 responsive RTP801 (RTP801), Protein regulated in development and DNA damage response 1, REDD-1DNA damage-inducible transcript 4 protein, REDD1HIF-1 responsive protein RTP801, RTP801RP11-442H21.1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-232 of human DDIT4 (NP_061931.1). MPSLWDRFSSSSTSSSPSSLPRTPTPDRPPRSAWGSATREEGFDRSTSLESSDCESLDSSNSGFGPEEDTAYLDGVSLPDFELLSDPEDEHLCANLMQLLQESLAQARLGSRRPARLLMPSQLVSQVGKELLRLAYSEPCGLRGALLDVCVEQGKSCHSVGQLALDPSLVPTFQLTLVLRLDSRLWPKIQGLFSSANSPFLPGFSQSLTLSTGFRVIKKKLYSSEQLLIEEC
Purification Method Affinity purified
Quantity 0.02 mL
Regulatory Status RUO
Research Discipline Cancer, mTOR Pathway
Primary or Secondary Primary
Gene ID (Entrez) 54541
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.