Learn More
VACUUBRAND™ DCP 300 Vacuum Gauge
Brand:  VACUUBRAND™ 20683171
Additional Details : Weight : 0.01000kg
Product Code. 11931002
  • 980.00 EUR / Each
Estimated Shipment: 24-06-2024
Specifications
0.44 kg
144 x 124 x 114 mm
Large Illuminated
Vacuum Gauge
114 mm