missing translation for 'onlineSavingsMsg'
Learn More

DBT Monoclonal Antibody (OTI1G2), TrueMAB™, OriGene

Product Code. 17552332
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17552332 30 μL 30µL
17532332 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17552332 Supplier Origene Technologies Supplier No. TA504748S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

DBT Monoclonal antibody specifically detects DBT in Human samples. It is validated for Flow Cytometry, Western Blot

Specifications

Antigen DBT
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI1G2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene CRYZL1
Gene Accession No. O95825
Gene Alias 52 kDa mitochondrial autoantigen of primary biliary cirrhosis; alpha-keto acid dehydrogenase precursor; BCATE2; BCKAD E2; BCKAD E2 subunit; BCKADE2; BCKAD-E2; BCOADC-E2; Branched chain 2-oxo-acid dehydrogenase complex component E2; branched chain acyltransferase, E2 component; branched-chain alpha-keto acid dehydrogenase complex component E2; branched-chain alpha-ketoacid dehydrogenase, E2 subunit; component of branched chain keto acid dehydrogenase complex; D3Wsu60e; DBT; Dihydrolipoamide acetyltransferase component of branched-chain alpha-keto acid dehydrogenase complex; Dihydrolipoamide branched chain transacylase; dihydrolipoamide branched chain transacylase E2; dihydrolipoyl transacylase; dihydrolipoyllysine-residue (2-methylpropanoyl)transferase; E2; E2 component of branched chain alpha-keto acid dehydrogenase complex; E2b; hypothetical protein LOC541388; im:7147214; lipoamide acyltransferase component of branched-chain alpha-keto acid dehydrogenase complex, mitochondrial; lipoamide acyltransferase component of mitochondrial branched-chain alpha-keto acid dehydrogenase complex; MGC9061; mitochondrial branched chain alpha-keto acid dehydrogenase transacylase subunit (E2b); part of the BCKAD complex; que; transacylase precursor; zgc:103768
Host Species Mouse
Immunogen Full length human recombinant protein of human CRYZL1 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9946
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.