Learn More
Tocris Bioscience™ dBRD9-A
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Description
dBRD9-A is a potent BRD9 Degrader (PROTAC™). Selectively binds BRD9 and elicits near complete degradation of BRD9 at nanomolar concentrations. Inhibits growth of synovial sarcoma cells in vitro and tumor progression in a synovial sarcoma xenograft mouse model. PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
| Chemical Name or Material | 2-((2,6-Dimethoxy-4-(2-methyl-1-oxo-1,2-dihydro-2,7-naphthyridin-4-yl)benzyl)(methyl)amino)-N-(8-((2-(2,6-dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)amino)octyl)acetamide |
| CAS | 2170679-42-0 |
| Quantity | 5 mg |
| Target | Active Degraders |
| Molecular Formula | C42H49N7O8 |
| Purity | 0.98 |