Learn More
Davercin, MedChemExpress
Product Code. 16482616
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
2mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16482616 25 mg 25mg
16480026 2 mg 2mg
16455827 5 mg 5mg
16413098 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16482616 Supplier Medchem Express Supplier No. HY100584
 Restricted Item
Estimated Shipment: 30-11-2026
MedChemExpress Davercin (Erythromycin Cyclocarbonate), derivative of Erythromycin, which is active against Gram-positive and some Gram-negative microorganisms.
Davercin (Erythromycin Cyclocarbonate), derivative of Erythromycin, which is active against Gram-positive and some Gram-negative microorganisms.
Specifications
Chemical Name or Material Davercin
CAS 55224-05-0
Color White
Purity Grade Notes Research
Molecular Formula C38H65NO14
Quantity 25 mg
Synonym Erythromycin Cyclocarbonate
Solubility Information DMSO : ≥ 50 mg/mL (65.80 mM)
SMILES C[C@]([C@H](OC([C@@H]([C@H]([C@H](C)[C@H]1O[C@@](O[C@H](C)C[C@@H]2N(C)C)([H])[C@@H]2O)O[C@@](O[C@@H](C)[C@@H]3O)([H])C[C@@]3(C)OC)C)=O)CC)(O4)[C@]([C@H](C([C@@H](C[C@]1(O)C)C)=O)C)([H])OC4=O
Molecular Weight (g/mol) 759.92
Formula Weight 759.92
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation