missing translation for 'onlineSavingsMsg'
Learn More

Dasabuvir, TRC

Product Code. 30364092
Change view
Click to view available options
Quantity:
5 mg
25 mg
250 mg
Unit Size:
250mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30364092 25 mg 25mg
30357973 5 mg 5mg
30357275 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30364092 Supplier Toronto Research Chemicals Supplier No. TRCD19355025MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1132935-63-7
Molecular Formula C26 H27 N3 O5 S
Molecular Weight (g/mol) 493.57
Synonym Methanesulfonamide, N-[6-[5-(3,4-dihydro-2,4-dioxo-1(2H)-pyrimidinyl)-3-(1,1-dimethylethyl)-2-methoxyphenyl]-2-naphthalenyl]-, N-[6-[5-(3,4-Dihydro-2,4-dioxo-1(2H)-pyrimidinyl)-3-(1,1-dimethylethyl)-2-methoxyphenyl]-2-naphthalenyl]methanesulfonamide, ABT 333, Dasabuvir, N-[6-[3-tert-Butyl-5-(2,4-dioxo-3,4-dihydropyrimidin-1(2H)-yl)-2-methoxyphenyl]naphthalen-2-yl]methanesulfonamide
IUPAC Name N-[6-[3-tert-butyl-5-(2,4-dioxopyrimidin-1-yl)-2-methoxyphenyl]naphthalen-2-yl]methanesulfonamide
SMILES COc1c(cc(cc1C(C)(C)C)N2C=CC(=O)NC2=O)c3ccc4cc(NS(=O)(=O)C)ccc4c3

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C26H27N3O5S/c1-26(2,3)22-15-20(29-11-10-23(30)27-25(29)31)14-21(24(22)34-4)18-7-6-17-13-19(28-35(5,32)33)9-8-16(17)12-18/h6-15,28H,1-5H3,(H,27,30,31)
Formula Weight 493.1671
Chemical Name or Material Dasabuvir
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.