Learn More
Dapivirine, TRC
Product Code. 30360525
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30360525 5 mg 5mg
30352101 10 mg 10mg
30354279 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30360525 Supplier Toronto Research Chemicals Supplier No. TRCD1855005MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 244767-67-7
Molecular Formula C20 H19 N5
Molecular Weight (g/mol) 329.4
Synonym Benzonitrile, 4-[[4-[(2,4,6-trimethylphenyl)amino]-2-pyrimidinyl]amino]-, 4-[[4-[(2,4,6-Trimethylphenyl)amino]-2-pyrimidinyl]amino]benzonitrile, AIDS 105293, Dapivirine, R 147681, TMC 120, TMC 120R147681
IUPAC Name 4-[[4-(2,4,6-trimethylanilino)pyrimidin-2-yl]amino]benzonitrile
SMILES Cc1cc(C)c(Nc2ccnc(Nc3ccc(cc3)C#N)n2)c(C)c1
Specifications
Quantity 5 mg
InChI Formula InChI=1S/C20H19N5/c1-13-10-14(2)19(15(3)11-13)24-18-8-9-22-20(25-18)23-17-6-4-16(12-21)5-7-17/h4-11H,1-3H3,(H2,22,23,24,25)
Formula Weight 329.164
Chemical Name or Material Dapivirine