missing translation for 'onlineSavingsMsg'
Learn More

Dansylhydrazine, TRC

Product Code. 30004127
Change view
Click to view available options
Quantity:
250 mg
500 mg
Unit Size:
250mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004127 500 mg 500mg
30007926 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30004127 Supplier Toronto Research Chemicals Supplier No. TRCD179705500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33008-06-9
Molecular Formula C12H15N3O2S
Molecular Weight (g/mol) 265.33
Synonym 1-Dimethylaminonaphthalene-5-sulfonyl Hydrazide, 5-(Dimethylamino)naphthalene-1-sulfonyl Hydrazide, Dansyl Hydrazide;
IUPAC Name 5-(dimethylamino)naphthalene-1-sulfonohydrazide
SMILES CN(C)c1cccc2c(cccc12)S(=O)(=O)NN

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C12H15N3O2S/c1-15(2)11-7-3-6-10-9(11)5-4-8-12(10)18(16,17)14-13/h3-8,14H,13H2,1-2H3
Formula Weight 265.0885
Chemical Name or Material Dansylhydrazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.