missing translation for 'onlineSavingsMsg'
Learn More

Damascenone, TRC

Product Code. 30011845
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011845 10 mg 10mg
30009648 5 mg 5mg
30006718 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011845 Supplier Toronto Research Chemicals Supplier No. TRCD13450510MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 23696-85-7
Molecular Formula C13 H18 O
Molecular Weight (g/mol) 190.28
Synonym beta-Damascenone, 2-Buten-1-one, 1-(2,6,6-trimethyl-1,3-cyclohexadien-1-yl)-, 1-(2,6,6-Trimethyl-1,3-cyclohexadien-1-yl)-2-buten-1-one, 1-(2,6,6-Trimethyl-1,3-cyclohexadienyl)-2-buten-1-one, 1-Crotonoyl-2,6,6-trimethyl-1,3-cyclohexadiene, 2,6,6-Trimethyl-1-(2-butenoyl)-1,3-cyclohexadiene, 2,6,6-Trimethyl-1-crotonyl-1,3-cyclohexadiene
IUPAC Name 1-(2,6,6-trimethylcyclohexa-1,3-dien-1-yl)but-2-en-1-one
SMILES CC=CC(=O)C1=C(C)C=CCC1(C)C

Specifica

Quantity 10 mg
InChI Formula InChI=1S/C13H18O/c1-5-7-11(14)12-10(2)8-6-9-13(12,3)4/h5-8H,9H2,1-4H3
Formula Weight 190.1358
Chemical Name or Material 1-(2,6,6-Trimethyl-1,3-cyclohexadien-1-yl)-2-buten-1-one
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.