missing translation for 'onlineSavingsMsg'
Learn More

Damascenone, TRC

Product Code. 30009648
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009648 5 mg 5mg
30006718 1 mg 1mg
30011845 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009648 Supplier Toronto Research Chemicals Supplier No. TRCD1345055MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23696-85-7
Molecular Formula C13 H18 O
Molecular Weight (g/mol) 190.28
Synonym beta-Damascenone, 2-Buten-1-one, 1-(2,6,6-trimethyl-1,3-cyclohexadien-1-yl)-, 1-(2,6,6-Trimethyl-1,3-cyclohexadien-1-yl)-2-buten-1-one, 1-(2,6,6-Trimethyl-1,3-cyclohexadienyl)-2-buten-1-one, 1-Crotonoyl-2,6,6-trimethyl-1,3-cyclohexadiene, 2,6,6-Trimethyl-1-(2-butenoyl)-1,3-cyclohexadiene, 2,6,6-Trimethyl-1-crotonyl-1,3-cyclohexadiene
IUPAC Name 1-(2,6,6-trimethylcyclohexa-1,3-dien-1-yl)but-2-en-1-one
SMILES CC=CC(=O)C1=C(C)C=CCC1(C)C

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C13H18O/c1-5-7-11(14)12-10(2)8-6-9-13(12,3)4/h5-8H,9H2,1-4H3
Formula Weight 190.1358
Chemical Name or Material 1-(2,6,6-Trimethyl-1,3-cyclohexadien-1-yl)-2-buten-1-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.