Learn More
Memmert™ DAkkS Factory Calibration for UN450plus
Product Code. 17580226 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
UN450plus Oven
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
17580226 UN450plus Oven Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17580226 Supplier Memmert™ Supplier No. UN450plusE39696
DAkkS factory calibration for a freely selectable temperature for Memmert Ovens.
  • For one free-selectable temperature value according to method C (DKD-R 5-7)
Specifications
For Use With (Equipment) UN450plus Oven
Product Type DAkkS Factory Calibration