Learn More
Memmert™ DAkkS Factory Calibration for UF450
Product Code. 17559345 Shop All Memmert Products
Change view
Click to view available options
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. unitSize
17559345 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17559345 Supplier Memmert™ Supplier No. UF450E39696
DAkkS factory calibration for a freely selectable temperature for Memmert Ovens.
  • For one free-selectable temperature value according to method C (DKD-R 5-7)
Specifications
For Use With (Equipment) UF450 Oven
Product Type DAkkS Factory Calibration