Learn More
Melford Daidzein
Product Code. 30018226
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30018226 25 mg 25mg
30018227 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30018226 Supplier Melford Supplier No. D200100.025
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
Inactive analog of Genistein.
Inactive analog of Genistein. Inhibits casein kinase II activity.
Specifications
CAS 486-66-8
Molecular Formula C15H10O4
Quantity 25 mg