missing translation for 'onlineSavingsMsg'
Learn More

Dacuronium Bromide, TRC

Product Code. 30134606
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134606 100 mg 100mg
30012048 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134606 Supplier Toronto Research Chemicals Supplier No. TRCD102700100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27115-86-2
Molecular Formula C33 H58 N2 O3 . 2 Br
Molecular Weight (g/mol) 690.63
Synonym Piperidinium, 1,1'-[(2β,3α,5α,16β,17β)-3-(acetyloxy)-17-hydroxyandrostane-2,16-diyl]bis[1-methyl-, dibromide (9CI), Piperidinium, 1,1'-(3α,17β-dihydroxy-5α-androstan-2β,16β-ylene)bis[1-methyl-, dibromide, 3-acetate (8CI), Androstane, piperidinium deriv., 1,1'-[(2β,3α,5α,16β,17β)-3-(Acetyloxy)-17-hydroxyandrostane-2,16-diyl]bis[1-methylpiperidinium]dibromide, Dacuronium bromide
IUPAC Name [(2S,3S,5S,8R,9S,10S,13S,14S,16S,17R)-17-hydroxy-10,13-dimethyl-2,16-bis(1-methylpiperidin-1-ium-1-yl)-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-3-yl] acetate;dibromide
SMILES [Br-].[Br-].CC(=O)O[C@H]1C[C@@H]2CC[C@@H]3[C@H](CC[C@]4(C)[C@@H](O)[C@H](C[C@@H]34)[N+]5(C)CCCCC5)[C@@]2(C)C[C@@H]1[N+]6(C)CCCCC6

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C33H58N2O3.2BrH/c1-23(36)38-30-20-24-12-13-25-26(33(24,3)22-29(30)35(5)18-10-7-11-19-35)14-15-32(2)27(25)21-28(31(32)37)34(4)16-8-6-9-17-34;;/h24-31,37H,6-22H2,1-5H3;2*1H/q+2;;/p-2/t24-,25+,26-,27-,28-,29-,30-,31-,32-,33-;;/m0../s1
Formula Weight 688.2814
Chemical Name or Material 1,1'-(3alpha-acetoxy-17beta-hydroxy-5alpha-androstan-2beta,16beta-ylene) bis(1-methylpiperidinium) dibromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.