missing translation for 'onlineSavingsMsg'
Learn More

Dactolisib, MedChemExpress

Product Code. 16467846
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16467846 100 mg 100mg
16443997 50 mg 50mg
16441316 200 mg 200mg
16461916 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16467846 Supplier Medchem Express Supplier No. HY50673

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Dactolisib (BEZ235) is an orally active and dual pan-class I PI3K and mTOR kinase inhibitor with IC50s of 4 nM/5 nM/7 nM/75 nM, and 20.7 nM for p110α/p110γ/p110δ/p110β and mTOR, respectively. Dactolisib (BEZ235) inhibits both mTORC1 and mTORC2.

Dactolisib (BEZ235) is an orally active and dual pan-class I PI3K and mTOR kinase inhibitor with IC50s of 4 nM/5 nM/7 nM/75 nM, and 20.7 nM for p110α/p110γ/p110δ/p110β and mTOR, respectively. Dactolisib (BEZ235) inhibits both mTORC1 and mTORC2.

Specifications

Chemical Name or Material Dactolisib
CAS 915019-65-7
Color White
Purity Grade Notes Research
Molecular Formula C30H23N5O
Quantity 100 mg
Synonym BEZ235 NVP-BEZ235
Solubility Information 5% TFA : 8.33 mg/mL (17.74 mM; ultrasonic and warming and heat to 60°C) ∣DMSO : 5 mg/mL (10.65 mM; ultrasonic and warming and heat to 60°C) ∣DMF : 2 mg/mL (4.26 mM; Need ultrasonic)
SMILES CN(C1=C2C3=CC(C4=CC5=CC=CC=C5N=C4)=CC=C3N=C1)C(N2C6=CC=C(C=C6)C(C)(C#N)C)=O
Molecular Weight (g/mol) 469.54
Formula Weight 469.54
Percent Purity 96.41%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.