Learn More
Dacthal Monoacid, TRC
Product Code. 30008486
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008486 5 mg 5mg
30006907 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30008486 Supplier Toronto Research Chemicals Supplier No. TRCD1016505MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 887-54-7
Molecular Formula C9 H4 Cl4 O4
Molecular Weight (g/mol) 317.94
Synonym 1,4-Benzenedicarboxylic acid, 2,3,5,6-tetrachloro-, monomethyl ester (9CI), Terephthalic acid, tetrachloro-, methyl ester (7CI), Terephthalic acid, tetrachloro-, monomethyl ester (8CI), Chlorthal monomethyl, Dacthal monoacid, Methyl 2,3,5,6-tetrachloroterephthalate, Monomethyl 2,3,5,6-tetrachloroterephthalate, Monomethyl tetrachloroterephthalate, Tetrachloroterephthalic acid methyl ester
IUPAC Name 2,3,5,6-tetrachloro-4-methoxycarbonylbenzoic acid
SMILES COC(=O)c1c(Cl)c(Cl)c(C(=O)O)c(Cl)c1Cl
Specifications
Quantity 5 mg
InChI Formula InChI=1S/C9H4Cl4O4/c1-17-9(16)3-6(12)4(10)2(8(14)15)5(11)7(3)13/h1H3,(H,14,15)
Formula Weight 315.8864
Chemical Name or Material Dacthal monoacid