Learn More
D-Valinamide hydrochloride, TRC
Product Code. 30456588
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456588 10 g 10g
30454784 1 g 1g
30449076 5 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30456588 Supplier Toronto Research Chemicals Supplier No. TRCV09405010G
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemische Identifikatoren
CAS 133170-58-8
Molecular Formula C5H12N2O . (HCl)
Molecular Weight (g/mol) 152.62
Synonym (R)-2-Amino-3-methylbutanamide hydrochloride
IUPAC Name (2R)-2-amino-3-methylbutanamide;hydrochloride
SMILES Cl.CC(C)[C@@H](N)C(=O)N
Spezifikation
InChI Formula InChI=1S/C5H12N2O.ClH/c1-3(2)4(6)5(7)8;/h3-4H,6H2,1-2H3,(H2,7,8);1H/t4-;/m1./s1
Quantity 10 g
Formula Weight 152.0716
Shipping Condition Room Temperature
Chemical Name or Material D-Valinamide hydrochloride