Learn More
D-Phenyl-d5-alanine-2,3,3-d3-N-FMOC, CDN
Specifications
Specifications
| Chemical Name or Material | N-[(9 H-Fluoren-9-ylmethoxy)carbonyl]-D-phenylalanine-alpha,beta,beta,2,3,4,5,6-D8 |
| CAS | 2483831-22-5 |
| Molecular Formula | C24 D8 H13 N O4 |
| Quantity | 0.1 g |
| Synonym | N-[(9 H-Fluoren-9-ylmethoxy)carbonyl]-D-phenylalanine-α,β,β,2,3,4,5,6-d8, (2 R)-2,3,3-Trideuterio-2-(9 H-fluoren-9-ylmethoxycarbonylamino)-3-(2,3,4,5,6-pentadeuteriophenyl)propanoic acid, D-Phenyl-d5-alanine-2,3,3-d3-N-FMOC, N-[(9 H-Fluoren-9-ylmethoxy)carbonyl]-D-phenylalanine-α,β,β,2,3,4,5,6-D8, N-(9-Fluorenylmethoxycarbonyl)-D-phenylalanine-d8, N-Fmoc-D-phenylalanine-d8 |
| InChI Formula | InChI=1 S/C24H21NO4/c26-23(27)22(14-16-8-2-1-3-9-16)25-24(28)29-15-21-19-12-6-4-10-17(19)18-11-5-7-13-20(18)21/h1-13,21-22 H,14-15H2,(H,25,28)(H,26,27)/t22-/m1/s1/i1D,2 D,3 D,8 D,9 D,14D2,22 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(c([2 H])c1[2 H])C([2 H])([2 H])[C@@]([2 H])(NC(=O)OCC2c3ccccc3c4ccccc24)C(=O)O |
| IUPAC Name | (2 R)-2,3,3-trideuterio-2-(9 H-fluoren-9-ylmethoxycarbonylamino)-3-(2,3,4,5,6-pentadeuteriophenyl)propanoic acid |
| Molecular Weight (g/mol) | 395.4771 |
| Formula Weight | 395.1973 g/mol |
| Show More |