missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Mannosamine hydrochloride, 98%

Product Code. 15447629
Change view
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15447629 1 g 1g
15457629 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15447629 Supplier Thermo Scientific Chemicals Supplier No. J66316.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5505-63-5 | C6H14ClNO5 | 215.63 g/mol

D-Mannosamine hydrochloride is used in the synthesis of non-natural Mannac analogs for the expression of thiols on cell-surface sialic acids. It is also used to investigate and screening of N-acetyl-beta-hexosaminidase inhibitor libraries targeting osteoarthritis. It acts as a precursor of N-propanoyl mannosamine and other N-acyl precursors of sialic acid (N-acyl neuraminic acids). It is a pharmaceutical compound used in the development of mannosylated liposomes for bioadhesive oral drug delivery through M cells of Peyer's patches.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Mannosamine hydrochloride is used in the synthesis of non-natural Mannac analogs for the expression of thiols on cell-surface sialic acids. It is also used to investigate and screening of N-acetyl-beta-hexosaminidase inhibitor libraries targeting osteoarthritis. It acts as a precursor of N-propanoyl mannosamine and other N-acyl precursors of sialic acid (N-acyl neuraminic acids). It is a pharmaceutical compound used in the development of mannosylated liposomes for bioadhesive oral drug delivery through M cells of Peyer′s patches.

Solubility
Soluble in water and methanol.

Notes
Hygroscopic. Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5505-63-5
Molecular Formula C6H14ClNO5
Molecular Weight (g/mol) 215.63
MDL Number MFCD00064557
InChI Key CBOJBBMQJBVCMW-DEZHIRTDSA-N
Synonym 2-amino-2-deoxy-d-mannosehydrochloride
PubChem CID 133109008
IUPAC Name (2R,3S,4R,5S)-2-amino-3,4,5,6-tetrahydroxyhexanal;hydrochloride
SMILES C(C(C(C(C(C=O)N)O)O)O)O.Cl

Specifications

Melting Point 168°C (decomposition)
Color White
Quantity 1 g
Sensitivity Hygroscopic
Solubility Information Soluble in water and methanol.
Optical Rotation −4° (c=2 in Water)
Formula Weight 215.63
Percent Purity 98%
Physical Form Powder
Chemical Name or Material D-Mannosamine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.