Learn More
Tocris Bioscience™ D-Luciferin sodium salt
GREENER_CHOICE
Product Code. 18706009
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18706009 10 mg 10mg
18743988 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18706009 Supplier Tocris Bioscience™ Supplier No. 5427/10
Firefly luciferase substrate
D-Luciferin sodium salt is a firefly luciferase substrate (Km = ∽2 μM). Used in many standard bioluminescence imaging (BLI) techniques, including monitoring tumour growth, and investigating intracellular signaling activity in vitro and in vivo. Cell permeable. Emission maximum = 562 nm.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 10 mg
Molecular Formula C11H7N2NaO3S2
Product Type D-Luciferin sodium salt
Purity >99%
Molecular Weight (g/mol) 302.3
Inhibitors D-Luciferin sodium salt
For Research Use Only