Learn More
D,L-Tryptophan Methyl Ester Benzaldimine, TRC
Product Code. 30455268
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30455268 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30455268 Supplier Toronto Research Chemicals Supplier No. TRCT89485010G
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material L-Tryptophan Methyl Ester','Benzaldimine
CAS 19779-75-0
Molecular Formula C19 H18 N2 O2
Quantity 10 g
InChI Formula InChI=1S/C19H18N2O2/c1-23-19(22)18(20-12-14-7-3-2-4-8-14)11-15-13-21-17-10-6-5-9-16(15)17/h2-10,12-13,18,21H,11H2,1H3/b20-12+/t18-/m0/s1
SMILES COC(=O)[C@H](Cc1c[nH]c2ccccc12)\N=C\c3ccccc3
IUPAC Name methyl (2S)-2-[(E)-benzylideneamino]-3-(1H-indol-3-yl)propanoate
Molecular Weight (g/mol) 306.36
Formula Weight 306.1368