missing translation for 'onlineSavingsMsg'
Learn More

D,L-Tosylisopropylideneglycerol-d5, TRC

Product Code. 30455735
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30455735 10 mg 10mg
30452281 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30455735 Supplier Toronto Research Chemicals Supplier No. TRCT66497710MG
 Restricted Item
Estimated Shipment: 22-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material D,L-Tosylisopropylideneglycerol-d5
CAS 1330076-68-0
Molecular Formula C13H13D5S
Quantity 10 mg
Synonym 2,2-Dimethyl-1,3-dioxolane-4-methanol 4-(4-Methylbenzenesulfonate)-d5, 2,2-Dimethyl-1,3-dioxolane-4-methanol p-Toluenesulfonate-d5, (+/-)-1,2-(Isopropylidene)glycerol 3-(p-Toluenesulfonate)-d5, 1,2-Isopropylidenedioxy-3-tosyloxypropane-d5, Solketal Tosylate-d5, NSC 36147-d5
InChI Formula InChI=1S/C13H18O5S/c1-10-4-6-12(7-5-10)19(14,15)17-9-11-8-16-13(2,3)18-11/h4-7,11H,8-9H2,1-3H3/i8D2,9D2,11D
SMILES [2H]C([2H])(OS(=O)(=O)c1ccc(C)cc1)C2([2H])OC(C)(C)OC2([2H])[2H]
IUPAC Name [dideuterio-(4,5,5-trideuterio-2,2-dimethyl-1,3-dioxolan-4-yl)methyl] 4-methylbenzenesulfonate
Molecular Weight (g/mol) 291.37
Formula Weight 291.1189
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.